Gene Transcripts
- Gene Symbol
- CSNK1G3
- Entrez Gene ID
- 1456
- Gene Name
- casein kinase 1 gamma 3
- Gene Aliases
- CKI-gamma 3, CSNK1G3L
- Chromosome Location
- Chr. 5: 123512099 - 123617045 on Build GRCh38
- UniGene ID
- Hs.129206
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031812.3 |
NP_001026982.1
|
2, 3 |
890 |
|
NM_001044723.2 |
NP_001038188.1
|
1, 2 |
418 |
|
NM_001270572.1 |
NP_001257501.1
|
1, 2 |
418 |
|
NM_004384.4 |
NP_004375.2
|
1, 2 |
418 |
|
XM_005271891.1 |
XP_005271948.1
|
2, 3 |
890 |
|
XM_005271892.2 |
XP_005271949.1
|
2, 3 |
890 |
|
XM_005271894.2 |
XP_005271951.1
|
2, 3 |
890 |
|
XM_005271895.2 |
XP_005271952.1
|
2, 3 |
890 |
|
XM_005271896.1 |
XP_005271953.1
|
2, 3 |
890 |
|
XM_017009069.1 |
XP_016864558.1
|
3, 4 |
958 |
|
XM_017009070.1 |
XP_016864559.1
|
2, 3 |
890 |
|
GenBank mRNA |
AF049089.1 |
- |
1, 2 |
201 |
|
AF049090.1 |
- |
1, 2 |
201 |
|
AK289405.1 |
- |
1, 2 |
363 |
|
BC047567.1 |
- |
2, 3 |
542 |
4817058 |
View Full Product Details