Gene Transcripts

Gene Symbol
ITIH5
Entrez Gene ID
80760
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 5
Gene Aliases
ITI-HC5, PP14776
Chromosome Location
Chr. 10: 7559270 - 7666998 on Build GRCh38
UniGene ID
Hs.498586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001851.2 NP_001001851.1 6 785
NM_030569.6 NP_085046.5 6 785
NM_032817.5 NP_116206.4 2 120
XM_011519713.2 XP_011518015.1 6 785
XM_011519714.2 XP_011518016.1 6 785
GenBank mRNA AB075833.1 - 3 326
AK027375.1 - 6 722
AK027831.1 - 3 304
AK075381.1 - 6 758
AL833203.1 - 6 630
AY238437.1 - 6 681
AY358426.1 - 6 750
BC136439.1 - 6 758
CR627109.1 - 1 5219
DB079183.1 - 2 120

View Full Product Details