Gene Transcripts
- Gene Symbol
- SPX
- Entrez Gene ID
- 80763
- Gene Name
- spexin hormone
- Gene Aliases
- C12orf39
- Chromosome Location
- Chr. 12: 21526257 - 21532947 on Build GRCh38
- UniGene ID
- Hs.130692
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030572.3 |
NP_085049.1
|
5, 6 |
502 |
|
NR_135187.1 |
|
2, 3 |
433 |
|
NR_135188.1 |
|
2, 3 |
305 |
|
GenBank mRNA |
AK027273.1 |
- |
5, 6 |
340 |
|
AK075342.1 |
- |
5, 6 |
456 |
|
AK311147.1 |
- |
2, 3 |
203 |
|
AK311217.1 |
- |
2, 3 |
294 |
|
BC004336.1 |
- |
5, 6 |
437 |
3631700 |
DA071522.1 |
- |
2, 3 |
306 |
|
DA260291.1 |
- |
2, 3 |
433 |
|
DA380664.1 |
- |
5, 6 |
501 |
|
View Full Product Details