Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030768.2 | NP_110395.1 | 9 | 889 | |
XM_005246106.1 | XP_005246163.1 | 9 | 872 | ||
XM_006712784.1 | XP_006712847.1 | 9 | 924 | ||
XM_011511946.2 | XP_011510248.1 | 8 | 749 | ||
XM_017005056.1 | XP_016860545.1 | 9 | 748 | ||
XM_017005057.1 | XP_016860546.1 | 9 | 1262 | ||
XM_017005058.1 | XP_016860547.1 | 8 | 787 | ||
GenBank mRNA | AK001043.1 | - | 9 | 901 | |
AK055417.1 | - | 4 | 2107 | ||
AL136850.1 | - | 9 | 814 | ||
AY024365.1 | - | 9 | 860 | ||
BC006576.2 | - | 9 | 889 | 3604852 | |
BC035746.1 | - | 8 | 1284 | ||
CR533533.1 | - | 9 | 759 | ||
FJ491380.1 | - | 9 | 782 | ||
FJ491381.1 | - | 10 | 749 | ||
HQ658059.1 | - | 8 | 782 | ||
HQ658060.1 | - | 6 | 535 |