Gene Transcripts

Gene Symbol
LDOC1L
Entrez Gene ID
84247
Gene Name
leucine zipper down-regulated in cancer 1 like
Gene Aliases
Mar6, Mart6, dJ1033E15.2
Chromosome Location
Chr. 22: 44492570 - 44498125 on Build GRCh38
UniGene ID
Hs.744991
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032287.2 NP_115663.2 2 532
GenBank mRNA AK296479.1 - 2 534
BC030232.1 - 2 532 5240032
CR456439.1 - 1 224

View Full Product Details