Gene Transcripts
- Gene Symbol
- DCTN5
- Entrez Gene ID
- 84516
- Gene Name
- dynactin subunit 5
- Gene Aliases
- Chromosome Location
- Chr. 16: 23641366 - 23676207 on Build GRCh38
- UniGene ID
- Hs.435941
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199743.1 |
NP_001186672.1
|
3, 4 |
415 |
|
NM_032486.3 |
NP_115875.1
|
3, 4 |
415 |
|
NR_037573.1 |
|
4, 5 |
516 |
|
GenBank mRNA |
AK027387.1 |
- |
3, 4 |
278 |
|
AK292843.1 |
- |
3, 4 |
342 |
|
AK299631.1 |
- |
3, 4 |
312 |
|
BC004191.2 |
- |
3, 4 |
295 |
3505855 |
CD388246.1 |
- |
3, 4 |
334 |
|
DA310703.1 |
- |
3, 4 |
415 |
|
DB087566.1 |
- |
4, 5 |
372 |
|
View Full Product Details