Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001129889.2 | NP_001123361.1 | 2 | 866 | |
NM_001322184.1 | NP_001309113.1 | 3 | 442 | ||
NM_001322185.1 | NP_001309114.1 | 2 | 388 | ||
NM_001322186.1 | NP_001309115.1 | 4 | 657 | ||
NM_001922.4 | NP_001913.2 | 2 | 866 | ||
XM_011521049.1 | XP_011519351.1 | 4 | 1490 | ||
XM_017020401.1 | XP_016875890.1 | 2 | 869 | ||
GenBank mRNA | AJ000503.1 | - | 2 | 838 | |
AK293115.1 | - | 2 | 649 | ||
BC028311.1 | - | 2 | 713 | 4705935 | |
D17547.1 | - | 2 | 853 | ||
DQ902581.1 | - | 2 | 731 | ||
L18967.1 | - | 2 | 823 | ||
S69231.1 | - | 2 | 788 |