Gene Transcripts
- Gene Symbol
- NDUFAF2
- Entrez Gene ID
- 91942
- Gene Name
- NADH:ubiquinone oxidoreductase complex assembly factor 2
- Gene Aliases
- B17.2L, MMTN, NDUFA12L, mimitin
- Chromosome Location
- Chr. 5: 60945129 - 61153037 on Build GRCh38
- UniGene ID
- Hs.591757
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_174889.4 |
NP_777549.1
|
1, 2 |
263 |
|
GenBank mRNA |
AB183433.1 |
- |
1, 2 |
136 |
|
AF087990.1 |
- |
1, 2 |
232 |
|
AK026051.1 |
- |
1, 2 |
263 |
|
AK291296.1 |
- |
1, 2 |
226 |
|
BC001753.1 |
- |
1 |
185 |
3354845 |
BC022869.1 |
- |
1, 2 |
164 |
|
BC033965.1 |
- |
1, 2 |
179 |
5288040 |
KU178834.1 |
- |
1, 2 |
136 |
|
KU178835.1 |
- |
1, 2 |
136 |
|
KU178836.1 |
- |
1, 2 |
136 |
|
View Full Product Details