Gene Transcripts

Gene Symbol
PRDM6
Entrez Gene ID
93166
Gene Name
PR domain 6
Gene Aliases
PRISM
Chromosome Location
Chr. 5: 123089146 - 123194266 on Build GRCh38
UniGene ID
Hs.135118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136239.1 NP_001129711.1 3, 4 1294
XM_011543726.2 XP_011542028.1 2, 3 1357
GenBank mRNA AF272898.1 - 3, 4 944

View Full Product Details