Gene Transcripts
- Gene Symbol
- TPTE2
- Entrez Gene ID
- 93492
- Gene Name
- transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
- Gene Aliases
- TPIP
- Chromosome Location
- Chr. 13: 19422877 - 19561574 on Build GRCh38
- UniGene ID
- Hs.377488
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001141968.1 |
NP_001135440.1
|
16, 17 |
1379 |
|
NM_001271850.1 |
NP_001258779.1
|
4, 5 |
658 |
|
NM_130785.3 |
NP_570141.3
|
15, 16 |
1289 |
|
NM_199254.2 |
NP_954863.2
|
18, 19 |
1520 |
|
NR_073485.1 |
|
20, 21 |
1632 |
|
NR_073487.1 |
|
12, 13 |
1057 |
|
GenBank mRNA |
AJ421032.1 |
- |
15, 16 |
1289 |
|
AK301741.1 |
- |
20, 21 |
1632 |
|
AY219890.1 |
- |
18, 19 |
1520 |
|
BC128146.1 |
- |
16, 17 |
1361 |
40112547 |
BC128147.1 |
- |
12, 13 |
1039 |
|
FJ969729.1 |
- |
4, 5 |
661 |
|
View Full Product Details