Gene Transcripts

Gene Symbol
HSPA12B
Entrez Gene ID
116835
Gene Name
heat shock protein family A (Hsp70) member 12B
Gene Aliases
C20orf60
Chromosome Location
Chr. 20: 3732670 - 3753111 on Build GRCh38
UniGene ID
Hs.516854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197327.1 NP_001184256.1 6 624
NM_001318322.1 NP_001305251.1 5 526
NM_052970.4 NP_443202.3 6 624
XM_011529151.2 XP_011527453.1 3 322
XM_017027632.1 XP_016883121.1 6 539
XM_017027633.1 XP_016883122.1 6 523
XM_017027634.1 XP_016883123.1 6 523
XM_017027635.1 XP_016883124.1 6 567
GenBank mRNA AK056712.1 - 6 561
AK297120.1 - 4 417
AK301294.1 - 5 492
BC090857.1 - 5 485
BC110881.1 - 6 573 5725594
BC117284.1 - 6 522 40125835
BC126263.1 - 6 522 8991979
BC143932.1 - 6 522 9052450
CB990660.1 - 6 655

View Full Product Details