Gene Transcripts

Gene Symbol
MUCL1
Entrez Gene ID
118430
Gene Name
mucin like 1
Gene Aliases
SBEM
Chromosome Location
Chr. 12: 54854515 - 54858393 on Build GRCh38
UniGene ID
Hs.348419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_058173.2 NP_477521.1 4 416
GenBank mRNA AY359062.1 - 4 387

View Full Product Details