Gene Transcripts
- Gene Symbol
- PIK3AP1
- Entrez Gene ID
- 118788
- Gene Name
- phosphoinositide-3-kinase adaptor protein 1
- Gene Aliases
- BCAP
- Chromosome Location
- Chr. 10: 96593312 - 96720522 on Build GRCh38
- UniGene ID
- Hs.310456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152309.2 |
NP_689522.2
|
12, 13 |
2077 |
|
XM_005269498.1 |
XP_005269555.1
|
11, 12 |
1600 |
|
XM_005269499.1 |
XP_005269556.1
|
11, 12 |
1624 |
|
XM_011539248.1 |
XP_011537550.1
|
12, 13 |
2069 |
|
XM_011539249.1 |
XP_011537551.1
|
12, 13 |
2069 |
|
GenBank mRNA |
AK092883.1 |
- |
11, 12 |
1600 |
|
AK122584.1 |
- |
1, 2 |
2906 |
|
AK125635.1 |
- |
12, 13 |
2069 |
|
BC029917.1 |
- |
5, 6 |
928 |
5168493 |
BX648550.1 |
- |
12, 13 |
1973 |
|
View Full Product Details