Gene Transcripts

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
Chromosome Location
Chr. 2: 124025287 - 124921201 on Build GRCh38
UniGene ID
Hs.660653
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130773.3 NP_570129.1 3 654
XM_006712258.1 XP_006712321.1 3 654
XM_017003316.1 XP_016858805.1 3 654
GenBank mRNA AB077881.1 - 3 290
AK056528.1 - 3 654

View Full Product Details