Gene Transcripts

Gene Symbol
SLC9B2
Entrez Gene ID
133308
Gene Name
solute carrier family 9 member B2
Gene Aliases
NHA2, NHE10, NHEDC2
Chromosome Location
Chr. 4: 103018033 - 103077371 on Build GRCh38
UniGene ID
Hs.546482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300754.1 NP_001287683.1 11 1617
NM_001300756.1 NP_001287685.1 10 1846
NM_178833.5 NP_849155.2 12 2154
XM_006714085.2 XP_006714148.1 12 2153
XM_006714086.3 XP_006714149.1 11 1982
GenBank mRNA AA233228.1 - 2 238
AK172823.1 - 12 2154
AK297010.1 - 11 1528
AK299888.1 - 10 1405
BC047447.1 - 12 1684 5295722
BI962996.1 - 1 18

View Full Product Details