Gene Transcripts

Gene Symbol
C5orf58
Entrez Gene ID
133874
Gene Name
chromosome 5 open reading frame 58
Gene Aliases
Chromosome Location
Chr. 5: 170231730 - 170252569 on Build GRCh38
UniGene ID
Hs.519749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102609.2 NP_001096079.1 3, 4 280
NM_001305393.1 NP_001292322.1 3, 4 299
NM_001305394.1 NP_001292323.1 3, 4 288
NM_001305395.1 NP_001292324.1 4, 5 454
NM_001305396.1 NP_001292325.1 4, 5 443
NM_001305397.1 NP_001292326.1 4, 5 427
XM_011534432.2 XP_011532734.1 3, 4 751
XM_017009029.1 XP_016864518.1 4, 5 1641
XM_017009030.1 XP_016864519.1 4, 5 1613
XM_017009031.1 XP_016864520.1 3, 4 340
XM_017009032.1 XP_016864521.1 3, 4 264
XM_017009033.1 XP_016864522.1 3, 4 331
GenBank mRNA BC092511.1 - 2, 3 1260
BU561730.1 - 4, 5 390
BX100176.1 - 2, 3 200
HY000873.1 - 3, 4 366
HY001392.1 - 3, 4 350
HY005987.1 - 3, 4 299
HY016595.1 - 3, 4 205
HY038777.1 - 3, 4 280

View Full Product Details