Gene Transcripts
- Gene Symbol
- UBE2F
- Entrez Gene ID
- 140739
- Gene Name
- ubiquitin conjugating enzyme E2 F (putative)
- Gene Aliases
- NCE2
- Chromosome Location
- Chr. 2: 237966945 - 238042782 on Build GRCh38
- UniGene ID
- Hs.471785
- Species
- Homo sapiens
- Gene Symbol
- UBE2F-SCLY
- Entrez Gene ID
- 100533179
- Gene Name
- UBE2F-SCLY readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 2: 237966945 - 238099413 on Build GRCh38
- UniGene ID
- Hs.471785
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037904.1 |
|
2 |
310 |
|
View Full Product Details