Gene Transcripts

Gene Symbol
BPIFB4
Entrez Gene ID
149954
Gene Name
BPI fold containing family B member 4
Gene Aliases
C20orf186, LPLUNC4, RY2G5, dJ726C3.5
Chromosome Location
Chr. 20: 33081512 - 33111751 on Build GRCh38
UniGene ID
Hs.38961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182519.2 NP_872325.2 5 828
GenBank mRNA AF549190.1 - 4 696

View Full Product Details