Gene Transcripts
- Gene Symbol
- URB1
- Entrez Gene ID
- 9875
- Gene Name
- URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
- Gene Aliases
- C21orf108, NPA1
- Chromosome Location
- Chr. 21: 32311019 - 32393003 on Build GRCh38
- UniGene ID
- Hs.473611
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014825.2 |
NP_055640.2
|
39 |
9517 |
|
GenBank mRNA |
AF231919.1 |
- |
39 |
9355 |
|
View Full Product Details