Gene Transcripts
- Gene Symbol
- EVC
- Entrez Gene ID
- 2121
- Gene Name
- EvC ciliary complex subunit 1
- Gene Aliases
- DWF-1, EVC1, EVCL
- Chromosome Location
- Chr. 4: 5711197 - 5829043 on Build GRCh38
- UniGene ID
- Hs.646899
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306090.1 |
NP_001293019.1
|
1, 2 |
363 |
|
NM_001306092.1 |
NP_001293021.1
|
1, 2 |
363 |
|
NM_153717.2 |
NP_714928.1
|
1, 2 |
363 |
|
XM_006713865.3 |
XP_006713928.1
|
1, 2 |
360 |
|
XM_006713866.3 |
XP_006713929.1
|
1, 2 |
360 |
|
XM_011513419.2 |
XP_011511721.1
|
1, 2 |
357 |
|
XM_017007883.1 |
XP_016863372.1
|
1, 2 |
357 |
|
GenBank mRNA |
AF216184.1 |
- |
1, 2 |
363 |
|
AF216185.1 |
- |
1, 2 |
363 |
|
AF239742.1 |
- |
1, 2 |
179 |
|
BC085608.1 |
- |
1, 2 |
345 |
30523373 |
View Full Product Details