Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320289.1 | NP_001307218.1 | 6 | 976 | |
NM_001320290.1 | NP_001307219.1 | 6 | 713 | ||
NM_178033.1 | NP_828847.1 | 6 | 1010 | ||
XM_017000973.1 | XP_016856462.1 | 6 | 1041 | ||
GenBank mRNA | AK091806.1 | - | 6 | 714 | |
AK098065.1 | - | 6 | 1003 | ||
AK131355.1 | - | 6 | 976 | ||
AM040940.1 | - | 6 | 847 | ||
AY358537.1 | - | 6 | 993 | ||
BC028102.1 | - | 6 | 1010 | 5243005 |