Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318804.1 | NP_001305733.1 | 3 | 537 | |
NM_178864.3 | NP_849195.2 | 3 | 537 | ||
XM_017017537.1 | XP_016873026.1 | 5 | 1976 | ||
XM_017017539.1 | XP_016873028.1 | 3 | 537 | ||
GenBank mRNA | AB049469.1 | - | 3 | 462 | |
AK096253.1 | - | 3 | 537 | ||
BC105001.1 | - | 3 | 450 | 8144004 | |
BC105003.1 | - | 3 | 450 | 8144006 | |
BC143630.1 | - | 3 | 450 |