Gene Transcripts
- Gene Symbol
- F9
- Entrez Gene ID
- 2158
- Gene Name
- coagulation factor IX
- Gene Aliases
- F9 p22, FIX, HEMB, P19, PTC, THPH8
- Chromosome Location
- Chr. X: 139530720 - 139563459 on Build GRCh38
- UniGene ID
- Hs.522798
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000133.3 |
NP_000124.1
|
4, 5 |
415 |
|
GenBank mRNA |
A13997.1 |
- |
4, 5 |
387 |
|
AK292749.1 |
- |
4, 5 |
386 |
|
BC109214.1 |
- |
4, 5 |
415 |
40007636 |
BC109215.1 |
- |
4, 5 |
415 |
40007637 |
FR846238.1 |
- |
4, 5 |
386 |
|
FR846239.1 |
- |
4, 5 |
386 |
|
FR846240.1 |
- |
4, 5 |
386 |
|
J00136.1 |
- |
4, 5 |
386 |
|
J00137.1 |
- |
4, 5 |
387 |
|
M11309.1 |
- |
4, 5 |
386 |
|
M35672.1 |
- |
3, 4 |
281 |
|
View Full Product Details