Gene Transcripts

Gene Symbol
RFESD
Entrez Gene ID
317671
Gene Name
Rieske Fe-S domain containing
Gene Aliases
Chromosome Location
Chr. 5: 95646744 - 95658082 on Build GRCh38
UniGene ID
Hs.399758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001131065.1 NP_001124537.1 6 1778
NM_001131066.1 NP_001124538.1 6 623
NM_173362.3 NP_775498.1 5 1680
XM_011543360.2 XP_011541662.1 7 1666
XM_011543361.2 XP_011541663.1 5 558
XM_017009413.1 XP_016864902.1 6 790
GenBank mRNA BC024023.1 - 4 397
BC035110.2 - 5 1678 5262922
BC048979.1 - 5 1705 4838006
BI551847.1 - 6 626
BM263054.1 - 5 587

View Full Product Details