Gene Transcripts

Gene Symbol
NCCRP1
Entrez Gene ID
342897
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
FBXO50, NCCRP-1
Chromosome Location
Chr. 19: 39196964 - 39201882 on Build GRCh38
UniGene ID
Hs.726934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001414.1 NP_001001414.1 6 745
GenBank mRNA AK123941.1 - 6 745
BC067874.1 - 6 745 30348184
BC092493.1 - 6 748 30337042

View Full Product Details