Gene Transcripts
- Gene Symbol
- NUP43
- Entrez Gene ID
- 348995
- Gene Name
- nucleoporin 43
- Gene Aliases
- bA350J20.1, p42
- Chromosome Location
- Chr. 6: 149724315 - 149746572 on Build GRCh38
- UniGene ID
- Hs.510375
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198887.2 |
NP_942590.1
|
4, 5 |
589 |
|
NR_104456.1 |
|
4, 5 |
589 |
|
XM_005266960.4 |
XP_005267017.1
|
4, 5 |
539 |
|
XM_005266961.3 |
XP_005267018.1
|
4, 5 |
539 |
|
XM_005266962.3 |
XP_005267019.1
|
4, 5 |
539 |
|
XM_011535798.2 |
XP_011534100.1
|
4, 5 |
539 |
|
XM_011535799.2 |
XP_011534101.1
|
4, 5 |
539 |
|
GenBank mRNA |
AF514997.1 |
- |
4, 5 |
522 |
|
AK074311.1 |
- |
|
544 |
|
AK095994.1 |
- |
4, 5 |
545 |
|
AK304505.1 |
- |
3, 4 |
396 |
|
BC040121.1 |
- |
2, 3 |
234 |
|
BC065028.1 |
- |
4, 5 |
515 |
6042215 |
BX647097.1 |
- |
4, 5 |
539 |
|
CB989591.1 |
- |
4, 5 |
586 |
|
DC324759.1 |
- |
4, 5 |
589 |
|
View Full Product Details