Gene Transcripts

Gene Symbol
TBC1D26
Entrez Gene ID
353149
Gene Name
TBC1 domain family member 26
Gene Aliases
Chromosome Location
Chr. 17: 15732277 - 15744784 on Build GRCh38
UniGene ID
Hs.434123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178571.4 NP_848666.2 10, 11 913
GenBank mRNA AB449913.1 - 8, 9 663
BC047400.1 - 10, 11 907 5297645

View Full Product Details