Gene Transcripts

Gene Symbol
TMEM81
Entrez Gene ID
388730
Gene Name
transmembrane protein 81
Gene Aliases
HC3107, KVLA2788, UNQ2788
Chromosome Location
Chr. 1: 205083129 - 205084460 on Build GRCh38
UniGene ID
Hs.146928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203376.1 NP_976310.1 1 753
GenBank mRNA AK303432.1 - 1 810
AY359081.1 - 1 753
BC061592.1 - 1 696 6202253

View Full Product Details