Gene Transcripts
- Gene Symbol
- FLT3
- Entrez Gene ID
- 2322
- Gene Name
- fms related tyrosine kinase 3
- Gene Aliases
- CD135, FLK-2, FLK2, STK1
- Chromosome Location
- Chr. 13: 28003274 - 28100592 on Build GRCh38
- UniGene ID
- Hs.507590
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004119.2 |
NP_004110.2
|
6, 7 |
818 |
|
NR_130706.1 |
|
6, 7 |
818 |
|
XM_011535015.2 |
XP_011533317.1
|
6, 7 |
817 |
|
XM_011535017.2 |
XP_011533319.1
|
6, 7 |
720 |
|
XM_011535018.2 |
XP_011533320.1
|
3, 4 |
328 |
|
XM_017020487.1 |
XP_016875976.1
|
7, 8 |
891 |
|
XM_017020488.1 |
XP_016875977.1
|
5, 6 |
660 |
|
XM_017020489.1 |
XP_016875978.1
|
5, 6 |
679 |
|
GenBank mRNA |
BC036028.1 |
- |
6, 7 |
818 |
|
BC126350.1 |
- |
6, 7 |
802 |
8992066 |
BC144039.1 |
- |
6, 7 |
803 |
9052559 |
BC144040.1 |
- |
6, 7 |
803 |
9052560 |
U02687.1 |
- |
6, 7 |
793 |
|
Z26652.1 |
- |
6, 7 |
736 |
|
View Full Product Details