Gene Transcripts
- Gene Symbol
- HIST2H2BF
- Entrez Gene ID
- 440689
- Gene Name
- histone cluster 2, H2bf
- Gene Aliases
- Chromosome Location
- Chr. 1: 149782689 - 149812373 on Build GRCh38
- UniGene ID
- Hs.632451
- Species
- Homo sapiens
- Gene Symbol
- HIST2H2BA
- Entrez Gene ID
- 337875
- Gene Name
- histone cluster 2, H2ba (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 1: 121108441 - 121117251 on Build GRCh38
- UniGene ID
- Hs.742189
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027337.1 |
|
1 |
127 |
|
GenBank mRNA |
BC106916.2 |
- |
1 |
127 |
|
BC126904.1 |
- |
1 |
128 |
|
View Full Product Details