Gene Transcripts
- Gene Symbol
- HIST2H2BF
- Entrez Gene ID
- 440689
- Gene Name
- histone cluster 2, H2bf
- Gene Aliases
- Chromosome Location
- Chr. 1: 149782689 - 149812373 on Build GRCh38
- UniGene ID
- Hs.632451
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK289664.1 |
- |
1 |
767 |
|
BC110793.1 |
- |
1 |
753 |
5224812 |
View Full Product Details