Gene Transcripts

Gene Symbol
CASC15
Entrez Gene ID
401237
Gene Name
cancer susceptibility candidate 15 (non-protein coding)
Gene Aliases
LINC00340
Chromosome Location
Chr. 6: 21666444 - 22194400 on Build GRCh38
UniGene ID
Hs.712707
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015410.1 2 422
GenBank mRNA AK022865.1 - 1 48
AK026189.1 - 2 422
AK300802.1 - 2 451
AK310212.1 - 3 542

View Full Product Details