Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079512.3 | NP_001072980.1 | 8 | 1181 | |
NM_001318873.1 | NP_001305802.1 | 8 | 1632 | ||
XM_011522654.2 | XP_011520956.1 | 8 | 2104 | ||
XM_017023649.1 | XP_016879138.1 | 8 | 1666 | ||
XM_017023650.1 | XP_016879139.1 | 8 | 1866 | ||
XM_017023651.1 | XP_016879140.1 | 8 | 1294 | ||
GenBank mRNA | AK124106.1 | - | 8 | 1489 | |
AK294418.1 | - | 8 | 1172 | ||
AL833564.1 | - | 6 | 1069 |