Gene Transcripts

Gene Symbol
Cdk5
Entrez Gene ID
12568
Gene Name
cyclin-dependent kinase 5
Gene Aliases
AW048668, Crk6
Chromosome Location
Chr. 5: 24407597 - 24423591 on Build GRCm38
UniGene ID
Mm.298798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007668.3 NP_031694.1 2 122
GenBank mRNA AK051790.1 - 2 122
BC052007.1 - 2 96 5706932
D29678.1 - 2 131

View Full Product Details