Gene Transcripts

Gene Symbol
Col4a4
Entrez Gene ID
12829
Gene Name
collagen, type IV, alpha 4
Gene Aliases
E130010M05Rik, [a]4(IV)
Chromosome Location
Chr. 1: 82448419 - 82586849 on Build GRCm38
UniGene ID
Mm.40253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007735.2 NP_031761.1 46 4803
GenBank mRNA AF169388.1 - 46 4746
BC038814.1 - 6 563
Z35167.1 - 3 342

View Full Product Details