Gene Transcripts

Gene Symbol
Fcer1g
Entrez Gene ID
14127
Gene Name
Fc receptor, IgE, high affinity I, gamma polypeptide
Gene Aliases
AI573376, CD23, FcR-gamma, FcR[g], FcRgamma, Fce1g, FcepsilonRI, Ly-50
Chromosome Location
Chr. 1: 171229572 - 171234349 on Build GRCm38
UniGene ID
Mm.22673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010185.4 NP_034315.1 5 492
GenBank mRNA AK155600.1 - 5 449
BC034163.1 - 432 5065647
CK791068.1 - 5 429
J05020.1 - 5 417

View Full Product Details