Gene Transcripts

Gene Symbol
Fyn
Entrez Gene ID
14360
Gene Name
Fyn proto-oncogene
Gene Aliases
AI448320, AW552119
Chromosome Location
Chr. 10: 39369770 - 39565381 on Build GRCm38
UniGene ID
Mm.4848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122892.1 NP_001116364.1 12 1767
NM_001122893.1 NP_001116365.1 13 1846
NM_008054.2 NP_032080.2 13 1837
GenBank mRNA AK156584.1 - 13 1848
AK171646.1 - 13 1839
BC010554.1 - 12 1448
BC032149.1 - 12 1482 3602546
BC092217.1 - 12 1763 6403711
M27266.1 - 14 1549
U70324.1 - 13 1550

View Full Product Details