Gene Transcripts

Gene Symbol
Gata4
Entrez Gene ID
14463
Gene Name
GATA binding protein 4
Gene Aliases
Gata-4
Chromosome Location
Chr. 14: 63198915 - 63271714 on Build GRCm38
UniGene ID
Mm.247669
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008092.3 NP_032118.2 5 1562
GenBank mRNA AB075549.1 - 5 1580
AF179424.1 - 4 983
AK134639.1 - 4 586
BC137824.1 - 4 1315 8860843
M98339.1 - 5 1530
U85046.1 - 5 1533

View Full Product Details