Gene Transcripts

Gene Symbol
Gng10
Entrez Gene ID
14700
Gene Name
guanine nucleotide binding protein (G protein), gamma 10
Gene Aliases
AV006524
Chromosome Location
Chr. 4: 59035156 - 59041899 on Build GRCm38
UniGene ID
Mm.41780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025277.3 NP_079553.1 3 619
GenBank mRNA AK003187.1 - 3 557
AK014158.1 - 3 579
BC061090.1 - 3 542

View Full Product Details