Gene Transcripts

Gene Symbol
Gypa
Entrez Gene ID
14934
Gene Name
glycophorin A
Gene Aliases
AI853584, CD235a, GPA
Chromosome Location
Chr. 8: 80493981 - 80510785 on Build GRCm38
UniGene ID
Mm.13123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010369.3 NP_034499.3 3 183
GenBank mRNA AK145998.1 - 3 184
AK157851.1 - 3 184
AK162614.1 - 3 168
BC148192.1 - 3 447 40045496

View Full Product Details