Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008321.2 | NP_032347.1 | 3 | 527 | |
GenBank mRNA | AK002820.1 | - | 3 | 525 | |
AK014306.1 | - | 1 | 167 | ||
AK031499.1 | - | 1143 | |||
AK201456.1 | - | 1 | 19 | ||
BC145871.1 | - | 3 | 527 | 40131174 | |
BC145873.1 | - | 3 | 527 | 40131176 | |
M60523.1 | - | 3 | 535 |