Gene Transcripts

Gene Symbol
Klra8
Entrez Gene ID
16639
Gene Name
killer cell lectin-like receptor, subfamily A, member 8
Gene Aliases
Cmv-1, Cmv1, Ly49h, Ly49u<129>
Chromosome Location
Chr. 6: 130115226 - 130129898 on Build GRCm38
UniGene ID
Mm.439698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101620.1 NP_001095090.1 3 338
NM_010650.3 NP_034780.1 266
GenBank mRNA AK143236.1 - 267
L78253.1 - 184
U12889.1 - 3 389

View Full Product Details