Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202485.1 | NP_001189414.1 | 2 | 596 | |
GenBank mRNA | BJ992310.1 | - | 2 | 214 | |
CR990871.1 | - | 2 | 180 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002157.2 | NP_002148.1 | 2 | 596 | |
GenBank mRNA | AK312104.1 | - | 2 | 192 | |
BC023518.2 | - | 2 | 270 | 4304224 | |
BG188453.1 | - | 1 | 157 | ||
CN264788.1 | - | 2 | 601 | ||
CR407688.1 | - | 1 | 156 | ||
U07550.1 | - | 2 | 197 | ||
X75821.1 | - | 1 | 156 |