Gene Transcripts

Gene Symbol
Nkx2-9
Entrez Gene ID
18094
Gene Name
NK2 homeobox 9
Gene Aliases
Nkx-2.9, Nkx2-8, Nkx2.9, tinman
Chromosome Location
Chr. 12: 56611389 - 56614676 on Build GRCm38
UniGene ID
Mm.129420
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008701.2 NP_032727.2 2 661
GenBank mRNA BC150762.1 - 2 645
Y15741.1 - 2 662

View Full Product Details