Gene Transcripts

Gene Symbol
Nppc
Entrez Gene ID
18159
Gene Name
natriuretic peptide type C
Gene Aliases
CNP, lbab
Chromosome Location
Chr. 1: 86666293 - 86670573 on Build GRCm38
UniGene ID
Mm.12802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010933.5 NP_035063.1 3 818
GenBank mRNA AK036176.1 - 3 822

View Full Product Details