Gene Transcripts

Gene Symbol
Olfr5
Entrez Gene ID
18349
Gene Name
olfactory receptor 5
Gene Aliases
MOR103-8, ORL120
Chromosome Location
Chr. 7: 6480216 - 6486909 on Build GRCm38
UniGene ID
Mm.358623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146914.2 NP_667125.1 2 580
GenBank mRNA BC104074.1 - 1 465
BC104075.1 - 1 475 40041745

View Full Product Details