Gene Transcripts

Gene Symbol
Osm
Entrez Gene ID
18413
Gene Name
oncostatin M
Gene Aliases
OncoM
Chromosome Location
Chr. 11: 4236785 - 4241026 on Build GRCm38
UniGene ID
Mm.131422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013365.2 NP_001013383.1 3 600
GenBank mRNA AK155637.1 - 3 551
BC099866.1 - 3 552 40040107

View Full Product Details