Gene Transcripts
- Gene Symbol
- Dpf2
- Entrez Gene ID
- 19708
- Gene Name
- D4, zinc and double PHD fingers family 2
- Gene Aliases
- 2210010M07Rik, BAF45D, Req, ubi-d4
- Chromosome Location
- Chr. 19: 5896516 - 5912871 on Build GRCm38
- UniGene ID
- Mm.2651
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291078.1 |
NP_001278007.1
|
4, 5 |
572 |
|
NM_011262.5 |
NP_035392.1
|
4, 5 |
572 |
|
GenBank mRNA |
AK004812.1 |
- |
4, 5 |
509 |
|
AK138047.1 |
- |
4, 5 |
506 |
|
AK144106.1 |
- |
4, 5 |
511 |
|
AK144848.1 |
- |
4, 5 |
507 |
|
AK144954.1 |
- |
4, 5 |
518 |
|
AK145851.1 |
- |
4, 5 |
568 |
|
AK156252.1 |
- |
3, 4 |
342 |
|
BC007188.1 |
- |
3, 4 |
360 |
|
BC012709.1 |
- |
4, 5 |
502 |
4208205 |
CX207082.1 |
- |
4, 5 |
496 |
|
U10435.1 |
- |
4, 5 |
459 |
|
U43921.1 |
- |
4, 5 |
524 |
|
View Full Product Details