Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277954.1 | NP_001264883.1 | 3, 4 | 374 | |
NM_001277955.1 | NP_001264884.1 | 3, 4 | 374 | ||
NM_017400.6 | NP_059096.3 | 4 | 381 | ||
GenBank mRNA | AK012114.1 | - | 316 | ||
BC027096.1 | - | 381 | 5254139 | ||
BC100453.1 | - | 219 | 6770826 | ||
BE653420.1 | - | 233 | |||
U58887.1 | - | 361 |