Gene Transcripts
- Gene Symbol
- Siah1b
- Entrez Gene ID
- 20438
- Gene Name
- seven in absentia 1B
- Gene Aliases
- AA960570, Sinh1b, siah-1b
- Chromosome Location
- Chr. X: 164070703 - 164076664 on Build GRCm38
- UniGene ID
- Mm.37215
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC052887.1 |
- |
4 |
1049 |
|
Z19580.1 |
- |
4 |
1054 |
|
View Full Product Details